missing translation for 'onlineSavingsMsg'
Learn More

S1P4/EDG-6 Antibody, PerCP, Novus Biologicals™

Product Code. 18402182
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18402182 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18402182 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP224500PCP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

S1P4/EDG-6 Polyclonal antibody specifically detects S1P4/EDG-6 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen S1P4/EDG-6
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias EDG6S1P receptor Edg-6, Endothelial differentiation G-protein coupled receptor 6, endothelial differentiation, G-protein-coupled receptor 6, endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor6,endothelial differentiation, G protein coupled receptor 6, S1P receptor 4, S1P4LPC1, SLP4, sphingosine 1-phosphate receptor 4, Sphingosine 1-phosphate receptor Edg-6, sphingosine-1-phosphate receptor 4
Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human Sphingolipid Receptor Edg6/S1P4 was used as the immunogen.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, GPCR, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8698
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.