missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ S1P3/EDG-3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £608.00
Specifications
| Antigen | S1P3/EDG-3 |
|---|---|
| Dilution | Western Blot : 5-10 μg/mL, Immunoprecipitation |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30066869
|
Novus Biologicals™
NBP3-30849-25ug |
25 μg |
£162.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30066952
|
Novus Biologicals™
NBP3-30849-100ug |
100 μg |
£608.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
S1P3/EDG-3 Polyclonal antibody specifically detects S1P3/EDG-3 in Human,Mouse samples. It is validated for Immunoprecipitation,Western BlotSpecifications
| S1P3/EDG-3 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cell Cycle and Replication, Cellular Markers, GPCR, Lipid and Metabolism, Signal Transduction | |
| Phosphate buffered saline | |
| 1903 | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Western Blot : 5-10 μg/mL, Immunoprecipitation | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| EDG-3, EDG3FLJ37523, Endothelial differentiation G-protein coupled receptor 3, endothelial differentiation, sphingolipid G-protein-coupled receptor, 3, FLJ93220, G protein-coupled receptor, endothelial differentiation gene-3, LPB3, MGC71696, S1P receptor 3, S1P receptor Edg-3, S1P receptor EDG3, S1P3, sphingosine 1-phosphate receptor 3, Sphingosine 1-phosphate receptor Edg-3, sphingosine-1-phosphate receptor 3 | |
| Rabbits were immunized with a synthetic peptide derived from the N terminal of the S1P3/EDG-3 protein. | |
| Ammonium sulfate precipitation |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title