missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S1P1/EDG-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
£169.00
Specifications
| Antigen | S1P1/EDG-1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 2 μg/mL, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500 |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
S1P1/EDG-1 Polyclonal specifically detects S1P1/EDG-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| S1P1/EDG-1 | |
| Western Blot 1 μg/mL, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 2 μg/mL, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| O08530 | |
| 1901 | |
| Synthetic peptide corresponding to residues S(359) H P Q K D D G D N P E T I(372) of mouse S1P1. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS containing 1mg/ml BSA with 0.05% Sodium Azide | |
| CD363, CD363 antigen, CHEDG1, D1S3362, EDG-1, EDG1edg-1, Endothelial differentiation G-protein coupled receptor 1, endothelial differentiation, sphingolipid G-protein-coupled receptor, 1, FLJ58121, S1P receptor 1, S1P receptor Edg-1, s1p1, S1P1ECGF1, sphingosine 1-phosphate receptor 1, sphingosine 1-phosphate receptor EDG1, Sphingosine 1-phosphate receptor Edg-1, sphingosine-1-phosphate receptor 1 | |
| S1PR1 | |
| IgG | |
| Affinity Purified | |
| NB120-11424 detects S1P1 protein (EDG1) in transfected human cell samples. This shows no cross-reactivity to S1P3. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title