missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S1P1/EDG-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB120-11424
This item is not returnable.
View return policy
Description
S1P1/EDG-1 Polyclonal specifically detects S1P1/EDG-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| S1P1/EDG-1 | |
| Polyclonal | |
| Unconjugated | |
| PBS containing 1mg/ml BSA with 0.05% Sodium Azide | |
| CD363, CD363 antigen, CHEDG1, D1S3362, EDG-1, EDG1edg-1, Endothelial differentiation G-protein coupled receptor 1, endothelial differentiation, sphingolipid G-protein-coupled receptor, 1, FLJ58121, S1P receptor 1, S1P receptor Edg-1, s1p1, S1P1ECGF1, sphingosine 1-phosphate receptor 1, sphingosine 1-phosphate receptor EDG1, Sphingosine 1-phosphate receptor Edg-1, sphingosine-1-phosphate receptor 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 1901 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1 ug/ml, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 2 ug/mL, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500 | |
| O08530 | |
| S1PR1 | |
| Synthetic peptide corresponding to residues S(359) H P Q K D D G D N P E T I(372) of mouse S1P1. | |
| 100 μg | |
| Primary | |
| NB120-11424 detects S1P1 protein (EDG1) in transfected human cell samples. This shows no cross-reactivity to S1P3. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction