missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S1P1/EDG-1 Antibody (713412) [Janelia Fluor™ 646], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB7089J
This item is not returnable.
View return policy
Description
S1P1/EDG-1 Monoclonal antibody specifically detects S1P1/EDG-1 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF
Specifications
| S1P1/EDG-1 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 713412 | |
| Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
| CD363, CD363 antigen, CHEDG1, D1S3362, EDG-1, EDG1edg-1, Endothelial differentiation G-protein coupled receptor 1, endothelial differentiation, sphingolipid G-protein-coupled receptor, 1, FLJ58121, S1P receptor 1, S1P receptor Edg-1, s1p1, S1P1ECGF1, sphingosine 1-phosphate receptor 1, sphingosine 1-phosphate receptor EDG1, Sphingosine 1-phosphate receptor Edg-1, sphingosine-1-phosphate receptor 1 | |
| Mouse S1P1/EDG-1 synthetic peptide (T4-H28), Accession # O08530 | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers, GPCR, Lipid and Metabolism, Neuroscience, Signal Transduction | |
| 1901 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction