missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ S100Z Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101793
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total S100Z.
Members of the S100 protein family contain 2 calcium-binding EF-hands and exhibit cell-type specific expression patterns. For additional background information on S100 proteins, see MIM 114085.
Specifications
| S100Z | |
| Polyclonal | |
| Unconjugated | |
| S100Z | |
| Gm625; Protein S100-Z; S100 calcium binding protein Z; S100 calcium binding protein, zeta; S100 calcium-binding protein Z; S100Z; S100-zeta | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 170591 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8WXG8 | |
| S100Z | |
| A synthesized peptide derived from human S100Z(Accession Q8WXG8), corresponding to amino acid residues E6-D56. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction