missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100P Rabbit anti-Human, Clone: S100P/4386R, Novus Biologicals™
Rabbit Monoclonal Antibody
£412.00
Specifications
| Antigen | S100P |
|---|---|
| Clone | S100P/4386R |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
Description
S100P Monoclonal specifically detects S100P in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| S100P | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| MIG9, migration-inducing gene 9, Protein S100-E, S100 calcium binding protein P, S100 calcium-binding protein Pprotein S100-P, S100E | |
| Recombinant fragment corresponding to C-terminal of human S100P protein | |
| Primary | |
| Store at 4°C. |
| S100P/4386R | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 6286 | |
| IgG | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title