missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ S100P Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103053
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total S100P.
The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21; however, this gene is located at 4p16. This protein, in addition to binding Ca2+, also binds Zn2+ and Mg2+. This protein may play a role in the etiology of prostate cancer.
Specifications
| S100P | |
| Polyclonal | |
| Unconjugated | |
| S100P | |
| MIG9; Migration-inducing gene 9 protein; protein S100-E; Protein S100-P; S100 calcium binding protein P; S100 calcium-binding protein P; S100E; S100p | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 6286 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P25815 | |
| S100P | |
| A synthesized peptide derived from human S100P(Accession P25815), corresponding to amino acid residues K30-A80. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction