Learn More
S100P Mouse anti-Human, Unconjugated, Clone: 1G6C3, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67485-1-IG-20UL
 View more versions of this product
Product Code. 16800146
  • £135.00 / 20µL
Description
The S100P antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human S100P. This antibody recognizes human antigen. The S100P antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
S100P
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, ELISA
1G6C3
Unconjugated
20 μL
6286
IgG2b