missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100P Antibody (S100P/9135R), Novus Biologicals™
Rabbit Monoclonal Antibody
£214.00 - £377.00
Specifications
| Antigen | S100P |
|---|---|
| Clone | S100P/9135R |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18691784
|
Novus Biologicals
NBP3-23858-20ug |
20 μg |
£214.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18656194
|
Novus Biologicals
NBP3-23858-100ug |
100 μg |
£377.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
S100P Monoclonal antibody specifically detects S100P in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| S100P | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| 10 mM PBS with 0.05% BSA | |
| 6286 | |
| IgG κ | |
| Protein A or G purified |
| S100P/9135R | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| MIG9, migration-inducing gene 9, Protein S100-E, S100 calcium binding protein P, S100 calcium-binding protein Pprotein S100-P, S100E | |
| Recombinant fragment (around aa1-95) of human S100P protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title