missing translation for 'onlineSavingsMsg'
Learn More

S100P Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£169.00 - £255.00

Specifications

Antigen S100P
Dilution Immunocytochemistry/ Immunofluorescence 1:300-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18604595
View Documents
Novus Biologicals
NBP2-99514-100ul
100 μg
£255.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18633476
View Documents
Novus Biologicals
NBP2-99514-50ul
50 μg
£169.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

S100P Polyclonal antibody specifically detects S100P in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications

Specifications

S100P
Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cancer
0.2 um filtered solution in PBS
6286
IgG
Antigen and protein A Affinity-purified
Immunocytochemistry/ Immunofluorescence 1:300-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000
Polyclonal
Purified
RUO
Human
MIG9, migration-inducing gene 9, Protein S100-E, S100 calcium binding protein P, S100 calcium-binding protein Pprotein S100-P, S100E
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human S100P.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documentos

Documentos

Certificados
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado