missing translation for 'onlineSavingsMsg'
Learn More

S100P Antibody (357517) [Allophycocyanin], Novus Biologicals™

Product Code. 30733330 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30733330 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733330 Supplier Novus Biologicals Supplier No. FAB2957A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

S100P Monoclonal antibody specifically detects S100P in Human samples. It is validated for Western Blot,Immunocytochemistry,Gene Knock-Out
TRUSTED_SUSTAINABILITY

Specifications

Antigen S100P
Applications Western Blot, Immunocytochemistry, Gene Knock-Out
Classification Monoclonal
Clone 357517
Conjugate APC
Formulation PBS
Gene Alias MIG9, migration-inducing gene 9, Protein S100-E, S100 calcium binding protein P, S100 calcium-binding protein Pprotein S100-P, S100E
Host Species Mouse
Immunogen E. coli-derived recombinant human S100P, Met1-Lys95, Accession # P25815
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 6286
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.