missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ S100G Polyclonal Antibody
GREENER_CHOICE

Product Code. 15813174
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15813174 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15813174 Supplier Invitrogen™ Supplier No. PA568289

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Predicted to react with Mouse and Rat samples.

S100G encodes calbindin D9K, a vitamin D-dependent calcium-binding protein. This cytosolic protein belongs to a family of calcium-binding proteins that includes calmodulin, parvalbumin, troponin C, and S100 protein. In the intestine, the protein is vitamin D-dependent and its expression correlates with calcium transport activity. The protein may increase Ca2+ absorption by buffering Ca2+ in the cytoplasm and increase ATP-dependent Ca2+ transport in duodenal basolateral membrane vesicles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen S100G
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 30% glycerol, 0.87% NaCl and 0.01% sodium azide; pH 7.3
Gene S100G
Gene Accession No. P02634, P29377, P97816
Gene Alias 9 kDa CaBP; AU042539; CABP; CABP1; CABP9K; CaBP-D9K; Calb3; calbindin 3, (vitamin D-dependent calcium binding protein); calbindin 3, (vitamin D-dependent calcium-binding protein); calbindin D9k; calbindin-D9K; calbindin-D9K major form; calcium-binding protein, intestinal, vitamin D-dependent (9-kDa CaBP); Cbpi; cholecalcin; cytidine 5'-triphosphate synthase 2; protein S100-G; Rncalbd9; S100 calcium binding protein G; S100 calcium-binding protein G; S100D; S100g; Vitamin D-dependent calcium-binding protein, intestinal
Gene Symbols S100G
Host Species Rabbit
Immunogen Synthesized peptide derived from human S100G.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12309, 24249, 795
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.