missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ S100B Monoclonal Antibody (CL2720)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA534997
This item is not returnable.
View return policy
Description
Immunogen sequence: LEKAMVALID VFHQYSGREG DKHKLKKSEL KELINNELSH FLEEIKEQEV VDKVMETLDN DGDGECDFQE FMAFVAMVTT ACHEFFEHE Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 98%.
S100B is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells and are involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100B may function in neurite extension, proliferation of melanoma cells, stimulation of Ca2+ fluxes, inhibition of PKC-mediated phosphorylation, astrocytosis and axonal proliferation, and inhibition of microtubule assembly. Chromosomal rearrangements and altered expression of this gene have been implicated in several diseases, including Alzheimer's disease, Down's syndrome, epilepsy, amyotrophic lateral sclerosis, melanoma, and type I diabetes.
Specifications
| S100B | |
| Monoclonal | |
| 1.0 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P04271, P04631, P50114 | |
| S100B | |
| Recombinant protein fragment of Human S100B (Product #RP-91304) | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| CL2720 | |
| Unconjugated | |
| S100B | |
| AI850290; Bpb; hypothetical protein LOC525716; NEF; protein S100-B; S100; S100 calcium binding protein B; S100 calcium binding protein beta (neural); S100 calcium-binding protein B; S100 calcium-binding protein beta (neural); S-100 calcium-binding protein beta subunit; S100 calcium-binding protein, beta (neural); S-100 calcium-binding protein, beta chain; S100 protein beta chain; S-100 protein beta chain; S-100 protein subunit beta; S100 protein, beta polypeptide; S100 protein, beta polypeptide, neural; S100B; S100-B; S100beta; S100P | |
| Mouse | |
| Protein A | |
| RUO | |
| 20203, 25742, 6285 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction