missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100B Antibody (S100B/1706R), DyLight 680, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-54426FR
This item is not returnable.
View return policy
Description
S100B Monoclonal antibody specifically detects S100B in Human, Mouse, Rat, Bovine, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array
Specifications
| S100B | |
| Monoclonal | |
| DyLight 680 | |
| beta (neural), NEF, S100, S100 beta, S100 calcium binding protein B, S100 calcium-binding protein B, S100 calcium-binding protein, beta (neural), S-100 calcium-binding protein, beta chain, 10protein S100-B, S-100 protein beta chain, S-100 protein subunit beta, S100beta | |
| Recombinant full-length human S100B protein (Uniprot: P04271) | |
| 100 μg | |
| Alzheimers Research, Apoptosis, BiologicAll speciesy Active Proteins, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Epigenetics, Hypoxia, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Peroxisome Markers, Signal Transduction, Stem Cells | |
| 6285 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array | |
| S100B/1706R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Bovine, Canine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction