missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ S100A9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587070
This item is not returnable.
View return policy
Description
Purity is > 95% by SDS-PAGE.
S100A9 (MRP-14) is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in the inhibition of casein kinase and altered expression of this protein is associated with the disease cystic fibrosis.
Specifications
| S100A9 | |
| Polyclonal | |
| Unconjugated | |
| S100A9 | |
| 60B8AG; AW546964; BEE22; BM-009; Cagb; calgranulin B; calgranulin-B; Calprotectin L1H subunit; CFAG; CGLB; DKFZp686B04128; GAGB; intracellular calcium-binding protein (MRP14); L1AG; Leukocyte L1 complex heavy chain; LIAG; MAC387; Macrophage; MIF; Migration inhibitory factor-related protein 14; Monocyte; MRP14; MRP-14; MRP15; MRP16; myeloid-related protein 14; NIF; P14; protein S100-A9; S100 A9; S100 calcium binding protein A9; S100 calcium binding protein A9 (calgranulin B); S100 calcium-binding protein A9; S100 calcium-binding protein A9 (calgranulin B); S100a9 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20202, 6280, 94195 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P06702, P31725, P50116 | |
| S100A9 | |
| Synthetic peptide of human S100A9. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction