missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A8 Rabbit anti-Human, Clone: 216, Novus Biologicals™
Rabbit Monoclonal Antibody
£316.00 - £457.00
Specifications
| Antigen | S100A8 |
|---|---|
| Clone | 216 |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:500-1:5000 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18205547
|
Novus Biologicals
NBP2-89808-50UL |
50 μg |
£316.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18265167
|
Novus Biologicals
NBP2-89808-100UL |
100 μg |
£457.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
S100A8 Monoclonal specifically detects S100A8 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| S100A8 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:500-1:5000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 60B8AG, CAGACP-10, calgranulin A, calgranulin-A, Calprotectin L1L subunit, CFAGL1Ag, CGLA, Cystic fibrosis antigen, Leukocyte L1 complex light chain, MA387, MIF, Migration inhibitory factor-related protein 8, MRP-8, MRP8S100 calcium binding protein A8 (calgranulin A), NIF, P8, protein S100-A8, S100 calcium binding protein A8, S100 calcium-binding protein A8, S100 calcium-binding protein A8 (calgranulin A), Urinary stone protein band A | |
| Recombinant Human S100A8 Protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 216 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| 0.2 um filtered solution in PBS | |
| 6279 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title