Learn More
S100A8, clone: CF-145, eBioscience™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 14-9745-80
Code : NEW
Additional Details : Weight : 0.09500kg
Description
The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in the inhibition of casein kinase and as a cytokine. Altered expression of this protein is associated with the disease cystic fibrosis.
Description: The monoclonal antibody CF-145 recognizes human S100A8. S100A8, also known as migration inhibitory factor-related protein 8 (MRP-8) or Calgranulin-A, is a calcium binding protein and member of the S100 family of proteins. S100A8 is expressed by neutrophils, monocytes, macrophages, granulocytes, and epithelial cells during dermatoses, such as psoriasis. S100A8 forms a heterodimer with S100A9, which together is known as calprotectin. S100A8 has antimicrobial and pro-apoptotic properties and is a pro-inflammatory mediator in acute and chronic inflammation. Levels of S100A8 are elevated in patients with various inflammatory disorders such as cystic fibrosis, rheumatoid arthritis, and chronic bronchitis. Upregulation of S100A8 is also observed in many tumors, including skin, breast, colon, pancreatic, and ovarian cancers.Applications Reported: This CF-145 antibody has been reported for use in western blotting, and immunohistochemical staining of formalin-fixed paraffin embedded tissue sections.Applications Tested: This CF-145 antibody has been tested by immunohistochemistry of formalin-fixed paraffin embedded tissue using low pH antigen retrieval and can be used at less than or equal to 1 μg/mL. This CF-145 antibody has been tested by western blot of A375 cell lysate and can be used at less than or equal to 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.Specifications
S100A8 | |
Monoclonal | |
0.5mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P05109 | |
S100A8 | |
Affinity chromatography | |
RUO | |
6279 | |
Store at 2-8°C. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
CF-145 | |
Unconjugated | |
S100A8 | |
60B8Ag, AI323541, B8Ag, CFAg, CP-10, Caga, MRP8, p8 | |
Mouse | |
25 μg | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
For Research Use Only.