missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ S100A8 Monoclonal Antibody (C2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA541673
This item is not returnable.
View return policy
Description
S100A8 Monoclonal Antibody for Western Blot, ICC/IF
The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in the inhibition of casein kinase and as a cytokine. Altered expression of this protein is associated with the disease cystic fibrosis.
Specifications
| S100A8 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P05109 | |
| S100A8 | |
| Recombinant protein S100 Calcium Binding Protein A8. The antigen corresponds to amino acid range 1-93 of the target protein. | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| Western Blot, Immunocytochemistry | |
| C2 | |
| Unconjugated | |
| S100A8 | |
| 60B8AG; AI323541; B8Ag; CAGA; CAGB; calgranulin A; calgranulin B; calgranulin-A; Calgranulin-B; calprotectin L1H subunit; Calprotectin L1L subunit; CFAg; CGLA; CGLB; chemotactic cytokine CP-10; chemotactic S100 protein; CP-10; Cystic fibrosis antigen; L1Ag; leukocyte L1 complex heavy chain; Leukocyte L1 complex light chain; LIAG; MA387; MAC387; MIF; Migration inhibitory factor-related protein 14; migration inhibitory factor-related protein 8; MRP14; MRP-14; MRP8; MRP-8; NIF; P14; p8; Pro-inflammatory S100 cytokine; protein S100-A8; Protein S100-A9; S100 A8; S100 calcium binding protein A8; S100 calcium binding protein A8 (calgranulin A); S100 calcium binding protein A9; S100 calcium-binding protein A8; S100 calcium-binding protein A8 (calgranulin A); S100 calcium-binding protein A9; S100 calcium-binding protein A9 (calgranulin B); S100A8; S100A9; Urinary stone protein band A | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 6279 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction