missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A8/A9 (MRP-8/14) Mouse anti-Human, Alexa Fluor 647, Clone: 5.5, BD Pharmingen™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566010
This item is not returnable.
View return policy
Description
S100A8/A9 (MRP-8/14) Monoclonal antibody specifically detects S100A8/A9 (MRP-8/14) in Human samples. It is validated for Flow cytometry (Intracellular Staining)) .
Specifications
| S100A8/A9 (MRP-8/14) | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Mouse | |
| 50 Tests | |
| Cell Biology | |
| Human | |
| Purified |
| Flow Cytometry, Flow Cytometry (Intracellular Staining) | |
| 5.5 | |
| Vol. Per test: 5μL | |
| S100A8/S100A9; MRP-8/MRP-14; Calprotectin, CFAG; p8,14 | |
| Acute Monocytic Leukemia Cells and Monocytes | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor™ 647 under optimum conditions and unreacted Alexa Fluor™ 647 was removed. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Reseach Use Only.
Spot an opportunity for improvement?Share a Content Correction