missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A8/A9 Antibody (5.5), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50283B
This item is not returnable.
View return policy
Description
S100A8/A9 Monoclonal specifically detects S100A8/A9 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| S100A8/A9 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| S100A8 | |
| Acute monocytic leukaemia cells. | |
| 0.1 mL | |
| Cancer | |
| 6279 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
| 5.5 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
| 60B8AG, CAGA, CFAG, CGLA, CP-10, L1Ag, MA387, MIF, MRP8, NIF, P8, S100 calcium binding protein A8, S100A8 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction