missing translation for 'onlineSavingsMsg'
Learn More

S100A7/Psoriasin Antibody (128), Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18336760 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18336760 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18336760 Supplier Novus Biologicals Supplier No. NBP289813AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

S100A7/Psoriasin Monoclonal specifically detects S100A7/Psoriasin in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen S100A7/Psoriasin
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone 128
Conjugate Alexa Fluor 700
Formulation 50 mM Sodium Borate
Gene Alias HID 5, HID5, HID-5, protein S100-A7, PSOR1, Psoriasin, psoriasin 1, S100 calcium binding protein A7, S100 calcium binding protein A7 (psoriasin 1), S100 calcium-binding protein A7, S100 calcium-binding protein A7 (psoriasin 1), S100A7c
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human S100A7/Psoriasin (NP_005219; Met668-Ala1210).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 6278
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.