missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A6 Mouse anti-Human, Clone: 3D11C8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66098-1-IG-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in stimulation of Ca2+-dependent insulin release, stimulation of prolactin secretion, and exocytosis. Chromosomal rearrangements and altered expression of this gene have been implicated in melanoma.Specifications
| S100A6 | |
| Monoclonal | |
| 0.6 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P06703 | |
| S100A6 | |
| S100A6 Fusion Protein Ag17606 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot | |
| 3D11C8 | |
| Unconjugated | |
| S100A6 | |
| 2A9, 5B10, CABP, CACY, Calcyclin, MLN 4, PRA, Protein S100 A6, S100A6 | |
| Mouse | |
| Protein G | |
| RUO | |
| 6277 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction