missing translation for 'onlineSavingsMsg'
Learn More

S100A6 Antibody (OTI5A10), DyLight 594, Novus Biologicals™

Product Code. 18411325 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18411325 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18411325 Supplier Novus Biologicals Supplier No. NBP273969DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

S100A6 Monoclonal antibody specifically detects S100A6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen S100A6
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI5A10
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias 2A9, CABP, CACY5B10, calcyclin, Growth factor-inducible protein 2A9, MLN 4, PRAS100 calcium binding protein A6 (calcyclin), Prolactin receptor-associated protein, protein S100-A6, S100 calcium binding protein A6, S100 calcium-binding protein A6, S100 calcium-binding protein A6 (calcyclin)
Host Species Mouse
Immunogen Full length recombinant protein of human S100A6 (NP_055439) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research
Primary or Secondary Primary
Gene ID (Entrez) 6277
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.