missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A5 Rabbit anti-Mouse, Clone: 1, Novus Biologicals™
Rabbit Monoclonal Antibody
£296.00 - £429.00
Specifications
| Antigen | S100A5 |
|---|---|
| Clone | 1 |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:20000-1:100000 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18255968
|
Novus Biologicals
NBP2-90581-50UL |
50 μg |
£296.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18233698
|
Novus Biologicals
NBP2-90581-100UL |
100 μg |
£429.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
S100A5 Monoclonal specifically detects S100A5 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| S100A5 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:20000-1:100000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 6276 | |
| IgG | |
| Protein A purified |
| 1 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| Protein S-100D, S100 calcium binding protein A5, S100 calcium-binding protein A5S100Dprotein S100-A5 | |
| Recombinant Mouse S100A5 protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title