missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ S100A5 Monoclonal Antibody (C8), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA543596
This item is not returnable.
View return policy
Description
Binds calcium, zinc and copper. One subunit can simultaneously bind 2 calcium ions or 2 copper ions plus 1 zinc ion. Calcium and copper ions compete for the same binding sites. Engelkamp D., Proc. Natl. Acad. Sci. U.S.A. 90:6547-6551(1993). Gregory S.G., Nature 441:315-321(2006).
Specifications
| S100A5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P33763 | |
| S100A5 | |
| Met1-Lys92 (Accession P33763), with N-terminal His Tag. | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| Immunohistochemistry (Paraffin), Western Blot | |
| C8 | |
| Unconjugated | |
| S100A5 | |
| Protein S100-A5; Protein S-100D; S100 calcium binding protein A5; S100 calcium-binding protein A4; S100 calcium-binding protein A5; S100a5; S100D; S100D9 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 6276 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction