missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98277-50ul
This item is not returnable.
View return policy
Description
S100A5 Polyclonal antibody specifically detects S100A5 in Mouse samples. It is validated for ELISA
Specifications
| S100A5 | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| Protein S-100D, S100 calcium binding protein A5, S100 calcium-binding protein A5S100Dprotein S100-A5 | |
| Produced in rabbits immunized with purified, recombinant Mouse S100A5 (Uniprot#: P63084; Met 1-Lys 93) | |
| 50 μg | |
| Neuroscience, Signal Transduction | |
| 6276 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction