missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16105-1-AP-20UL
This item is not returnable.
View return policy
Description
S100 belongs to the family of calcium binding proteins such as calmodulin and troponin C. S100A is composed of an alpha and beta chain whereas S100B is composed of two beta chains. S100 protein is also expressed in the antigen presenting cells such as the Langerhans cells in skin and interdigitating reticulum cells in the paracortex of lymph nodes.Specifications
| S100A4 | |
| Polyclonal | |
| Unconjugated | |
| S100A4 | |
| 18A2, 42A, Calvasculin, CAPL, Fibroblast Specific Protein, Fibroblast-specific protein 1, FSP1, FSP-1, Metastasin, MTS1, P9KA, PEL98, Protein Mts1, Protein S100 A4, S100A4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20198, 6275 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P07091, P26447 | |
| S100A4 | |
| S100A4 Fusion Protein Ag9019 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction