missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A4 Mouse, Clone: S100A4/1482, enQuireBio™
Mouse Monoclonal Antibody
£181.83 - £423.29
Specifications
| Antigen | S100A4 |
|---|---|
| Clone | S100A4/1482 |
| Concentration | 200μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
Specifications
| S100A4 | |
| 200μg/mL | |
| Monoclonal | |
| Mouse | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| 6275 | |
| Recombinant human S100A4 protein | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
| 10-12kDa |
| S100A4/1482 | |
| Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| NP_062427 | |
| S100A4 | |
| IgG1 κ | |
| Protein A/G Purified | |
| Cytoplasmic and Nuclear | |
| HeLa, A549 or A375 Cells. Placenta |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title