missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A4 Antibody (CL0237) [PE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52890PE
This item is not returnable.
View return policy
Description
S100A4 Monoclonal antibody specifically detects S100A4 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| S100A4 | |
| Monoclonal | |
| PE | |
| 18A2, Calvasculin, CAPLS100 calcium binding protein A4 (calcium protein, calvasculin, metastasin, fibroblast-specific protein-1,42A, FSP1, leukemia multidrug resistance associated protein, malignant transformation suppression 1, Metastasin, MTS1, murine placental homolog), P9KA, PEL98, Placental calcium-binding protein, Protein Mts1, protein S100-A4, S100 calcium binding protein A4, S100 calcium-binding protein A4, S100 calcium-binding protein A4 (calcium protein, calvasculin, metastasin, murine placental homolog) | |
| This antibody was developed using a recombinant protein derived from P26447, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer | |
| 6275 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL0237 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction