missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A3 Rabbit anti-Mouse, Clone: 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90583-50UL
This item is not returnable.
View return policy
Description
S100A3 Monoclonal specifically detects S100A3 in Mouse samples. It is validated for ELISA.
Specifications
| S100A3 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 6274 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| 1 | |
| ELISA 1:5000-1:10000 | |
| Protein S-100E, S100 calcium binding protein A3, S100 calcium-binding protein A3S100Eprotein S100-A3 | |
| Recombinant Mouse S100A3 Protein | |
| 50 μg | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction