Learn More
S100A3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12343-1-AP-20UL
 View more versions of this product
Product Code. 16810014
  • £135.00 / 20µL
Description
The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein has the highest content of cysteines of all S100 proteins, has a high affinity for Zinc, and is highly expressed in human hair cuticle. The precise function of this protein is unknown.
Specifications
Specifications
S100A3
Polyclonal
Unconjugated
S100A3
Protein S 100E, Protein S100 A3, S100A3, S100E
Rabbit
Antigen Affinity Chromatography
RUO
20197, 6274
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P33764, P62818
S100A3
S100A3 Fusion Protein Ag2995
20 μL
Primary
Mouse, Human