missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ S100A3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583623
This item is not returnable.
View return policy
Description
Immunogen sequence: KELATWTPTE FRECDYNKFM SVLDTNKDCE VDFVEYVRSL ACLCLYCHEY FKD Highest antigen sequence indentity to the following orthologs: Mouse - 91%, Rat - 91%.
The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein has the highest content of cysteines of all S100 proteins, has a high affinity for Zinc, and is highly expressed in human hair cuticle. The precise function of this protein is unknown.
Specifications
| S100A3 | |
| Polyclonal | |
| Unconjugated | |
| S100A3 | |
| Protein S100-A3; Protein S-100E; S100 calcium binding protein A3; S100 calcium-binding protein A3; S100A3; S100E | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6274 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P33764 | |
| S100A3 | |
| Recombinant protein corresponding to Human S100A3. Recombinant protein control fragment (Product #RP-99970). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction