missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A2 Antibody (CPTC-S100A2-2), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79899F
This item is not returnable.
View return policy
Description
S100A2 Monoclonal specifically detects S100A2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| S100A2 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| S100A2 | |
| Recombinant full-length human S100A2 protein (Uniprot: P29034) | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers, Neuroscience | |
| Human | |
| IgG2b κ |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| CPTC-S100A2-2 | |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| CAN19protein S100-A2, MGC111539, S100 calcium binding protein A2, S100 calcium-binding protein A2Protein S-100L, S100L | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction