missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10489-1-AP-20UL
This item is not returnable.
View return policy
Description
S100A14 gene encodes a member of the S100 protein family which contains an EF-hand motif and binds calcium. The gene is located in a cluster of S100 genes on chromosome 1. Levels of the encoded protein have been found to be lower in cancerous tissue and associated with metastasis suggesting a tumor suppressor function.Specifications
| S100A14 | |
| Polyclonal | |
| Unconjugated | |
| S100A14 | |
| Protein S100 A14, S100A14, S100A15, S114 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 57402, 66166, 685385 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin) | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9D2Q8, Q9HCY8 | |
| S100A14 | |
| S100A14 Fusion Protein Ag0757 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction