missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ S100A14 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106282
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total S100A14.
S100A14 gene encodes a member of the S100 protein family which contains an EF-hand motif and binds calcium. The gene is located in a cluster of S100 genes on chromosome 1. Levels of the encoded protein have been found to be lower in cancerous tissue and associated with metastasis suggesting a tumor suppressor function.
Specifications
| S100A14 | |
| Polyclonal | |
| Unconjugated | |
| S100a14 | |
| 1110013O05Rik; BCMP84; breast cancer membrane protein 84; Gm1020; Protein S100-A14; S100 calcium binding protein A14; S100 calcium binding protein A15; S100 calcium-binding protein A14; S100A14; S100a15; S114 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 57402, 685385 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9HCY8 | |
| S100a14 | |
| A synthesized peptide derived from human S100A14(Accession Q9HCY8), corresponding to amino acid residues M1-E35. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction