missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A13 Rabbit anti-Human, Clone: 20, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89734-50UL
This item is not returnable.
View return policy
Description
S100A13 Monoclonal specifically detects S100A13 in Human samples. It is validated for ELISA.
Specifications
| S100A13 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 6284 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| 20 | |
| ELISA 1:5000-1:10000 | |
| S100 calcium binding protein A13, S100 calcium-binding protein A13protein S100-A13 | |
| Recombinant Human S100A13 protein | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction