missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ S100A13 Antibody (S100A13/7483) [Alexa Fluor™ 750], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27004AF750
This item is not returnable.
View return policy
Description
S100A13 Monoclonal antibody specifically detects S100A13 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| S100A13 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| S100 calcium binding protein A13, S100 calcium-binding protein A13protein S100-A13 | |
| Recombinant fragment (around aa1-93) of human S100A13 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 6284 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Immunohistochemistry (Paraffin) | |
| S100A13/7483 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction