missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
S100A13 Polyclonal specifically detects S100A13 in Human samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | S100A13 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | DyLight 488 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | S100 calcium binding protein A13, S100 calcium-binding protein A13protein S100-A13 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant S100A13 (NP_001019381.1; Met 1-Lys 98). |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?