missing translation for 'onlineSavingsMsg'
Learn More

S100A13 Antibody (184), Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18408546 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18408546 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18408546 Supplier Novus Biologicals Supplier No. NBP289547AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

S100A13 Monoclonal antibody specifically detects S100A13 in Mouse samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen S100A13
Applications ELISA
Classification Monoclonal
Clone 184
Conjugate Alexa Fluor 594
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias S100 calcium binding protein A13, S100 calcium-binding protein A13protein S100-A13
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse S100A13 (P97352; Met1-Lys98).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 6284
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.