missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
S100A13 Monoclonal antibody specifically detects S100A13 in Human samples. It is validated for ELISA
Specifications
Specifications
| Antigen | S100A13 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 020 |
| Conjugate | DyLight 488 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | S100 calcium binding protein A13, S100 calcium-binding protein A13protein S100-A13 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human S100A13 (NP_001019381.1; Met1-Lys98). |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?