missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A11 Polyclonal Antibody

S100A11 Polyclonal Antibody
Brand: Invitrogen PA5102787
This item is not returnable.
View return policy
Description
S100A11 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
| S100A11 | |
| Polyclonal | |
| Unconjugated | |
| S100A11 | |
| cal; Calgizzarin; calizzarin; EMAP; Emap1; EMAPI; endothelial monocyte activating polypeptide 1; endothelial monocyte-activating polypeptide; epididymis secretory protein Li 43; HEL-S-43; Metastatic lymph node gene 70 protein; MLN 70; MLN70; Protein S100-A11; Protein S100-A11, N-terminally processed; Protein S100C; Protein S100-C; S100 calcium binding protein A11; S100 calcium binding protein A11 (calgizzarin); S100 calcium binding protein A11 (calizzarin); S100 calcium binding protein A14; S100 calcium-binding protein A11; S100 calcium-binding protein A11 (calgizzarin); S100a11; S100a14; S100C | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 20195, 445415, 6282 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P31949, P50543, Q6B345 | |
| S100A11 | |
| A synthesized peptide derived from human S100A11(Accession P31949), corresponding to amino acid residues G26-D76. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction