missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-13270
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
S100A11 Polyclonal specifically detects S100A11 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifica
| S100A11 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| S100A11 | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: SLIAVFQKYAGKDGYNYTLSKTEFLSFMNTELAAFTKNQKDPGVLDRMMKKLDTNSDGQLDFSEFLNLIGGLAMACHDSF | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 6282 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| 0.05mg/mL | |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| calgizzarin, Metastatic lymph node gene 70 protein, MLN 70, MLN70, protein S100-A11, Protein S100-C, S100 calcium binding protein A11, S100 calcium-binding protein A11, S100 calcium-binding protein A11 (calgizzarin), S100CS100 calcium binding protein A11 (calgizzarin) | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto