missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
S100A11 Polyclonal specifically detects S100A11 in Human samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | S100A11 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | FITC |
| Formulation | PBS |
| Gene Alias | calgizzarin, Metastatic lymph node gene 70 protein, MLN 70, MLN70, protein S100-A11, Protein S100-C, S100 calcium binding protein A11, S100 calcium-binding protein A11, S100 calcium-binding protein A11 (calgizzarin), S100CS100 calcium binding protein A11 (calgizzarin) |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant S100A11 (NP_005611.1; Met1-Thr105). |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?