missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ S100A10 Monoclonal Antibody (4E7E10)

Description
MA5-15326 targets S100A10/P11 in indirect ELISA, IF, IHC, and WB applications and shows reactivity with Human samples. The MA5-15326 immunogen is purified recombinant fragment of human P11 expressed in E. Coli. MA5-15326 detects S100A10/P11 which has a predicted molecular weight of approximately 11kDa.
The S-100 family of calcium-activated proteins interact with a range of target proteins to modulate biological signaling pathways. Numerous cancer cell lines overexpress the plasminogen receptor S-100A10 on the extracellular cell surface, where it forms a heterotetrameric complex with Annexin II, though this association is not required for plasma membrane localization or binding and activation of plasminogen. Additionally, S-100A10 acts as a cellular chaperone for hepatitis B (Hep B) virus polymerase. Hep B virus polymerase normally localizes to the cytoplasm only, though in the presence of S-100A10 a portion relocates to the nucleus, implying a role for S-100A10 and intracellular calcium in the process of viral replication.
Specifications
Specifications
| Antigen | S100A10 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 4E7E10 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Ascites with 0.03% sodium azide |
| Gene | S100A10 |
| Gene Accession No. | P60903 |
| Gene Alias | 42C; AA409961; AL024248; Annexin II; annexin II ligand; annexin II ligand, calpactin I, light polypeptide; annexin II tetramer (AIIt) p11 subunit; ANX2L; ANX2LG; Ca[1]; CAL12; Cal1l; calcium binding protein A11 (calgizzarin); calpactin I light chain; Calpactin-1 light chain; cellular ligand of annexin II; CLP11; GP11; MGC111133; MGC133268; Nerve growth factor-induced protein 42C; OTTHUMP00000015270; P PRSS26; p10; p10 protein; P11; p11 subunit; Protein S100 A10; Protein S100-A10; S100 calcium binding protein A10; S100 calcium binding protein A10 (annexin II ligand, calpactin I, light polypeptide (p11)); S100 calcium binding protein A10 (calgizzarin); S100 calcium binding protein A10 (calpactin); S100 calcium-binding protein A10; S100 calcium-binding protein A10 (annexin II ligand, calpactin I, light polypeptide (p11)); S-100 related protein, clone 42C; S100a10 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?