missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A10 Antibody (344015), PE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB2377P
This item is not returnable.
View return policy
Description
S100A10 Monoclonal antibody specifically detects S100A10 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
Specifications
| S100A10 | |
| Monoclonal | |
| PE | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
| 344015 | |
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| annexin II ligand, calpactin I, light polypeptide, ANX2L, ANX2LG, CAL1LGP11,42C, Calpactin I light chain, Calpactin-1 light chain, Cellular ligand of annexin II, CLP11Ca[1], MGC111133, p10, p10 protein, p11, protein S100-A10, S100 calcium binding protein A10, S100 calcium binding protein A10 (annexin II ligand, calpactin I, lightpolypeptide (p11)), S100 calcium-binding protein A10, S100 calcium-binding protein A10 (annexin II ligand, calpactin I, lightpolypeptide (p11)) | |
| E. coli-derived recombinant mouse S100A10, Met1-Lys97, Accession # P08207 | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction | |
| 6281 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction