missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A1 Antibody (D28-E), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP1-30154-500UL
This item is not returnable.
View return policy
Description
S100A1 Monoclonal specifically detects S100A1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| S100A/B | |
| Monoclonal | |
| Unconjugated | |
| 20mM Tris/HCl (pH 8.0) and 20 mg/ml BSA with 0.05% Sodium Azide | |
| protein S100-A1, S100 alpha, S100 calcium binding protein A1, S100 calcium-binding protein A1S100, S-100 protein alpha chain, S-100 protein subunit alpha, S100 protein, alpha polypeptide, S100A, S100-alpha | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody detects S100A and S100B monomers, the alpha-beta heterodimer, and also the beta-beta homodimer | |
| Store at 4C. Do not freeze. |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| D28-E | |
| Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:200, Immunohistochemistry-Frozen 1:100-1:200 | |
| P23297 | |
| S100A1 | |
| Peptide derived from C-terminal sequence of human S100A/B protein. | |
| 500 μL | |
| Astrocyte Markers, Breast Cancer, Cell Biology, Cytoskeleton Markers, Neuronal Cell Markers, Neuroscience | |
| 6271 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction