missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A1 Antibody (148), Alexa Fluor™ 488, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90505AF488
This item is not returnable.
View return policy
Description
S100A1 Monoclonal specifically detects S100A1 in Mouse samples. It is validated for ELISA, Sandwich ELISA Detection.
Specifications
| S100A1 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| protein S100-A1, S100 alpha, S100 calcium binding protein A1, S100 calcium-binding protein A1S100, S-100 protein alpha chain, S-100 protein subunit alpha, S100 protein, alpha polypeptide, S100A, S100-alpha | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse S100A1 (P56565; Gly2-Ser94). | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cell Biology, Cellular Markers, Hypoxia, Lipid and Metabolism, Neuronal Cell Markers, Neuroscience, Peroxisome Markers, Signal Transduction | |
| 6271 | |
| Mouse | |
| Purified |
| ELISA | |
| 148 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| No cross-reactivity in ELISA with: Mouse S100B | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction