missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A/B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£175.00 - £422.00
Specifications
| Antigen | S100A/B |
|---|---|
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Regulatory Status | RUO |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18233381
|
Novus Biologicals
NB200-538-0.1ML |
0.1 mL |
£175.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18179779
|
Novus Biologicals
NB200-538 |
0.5 mL |
£422.00
0.50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
S100A/B Polyclonal specifically detects S100A/B in Human, Mouse, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| S100A/B | |
| Unconjugated | |
| RUO | |
| P23297, P56565 | |
| 6271 | |
| Affinity purified S-100 protein isolated from cow brain. | |
| Primary | |
| Store at 4°C. |
| Polyclonal | |
| Rabbit | |
| Astrocyte Markers, Breast Cancer, Cell Biology, Cytoskeleton Markers, Neuronal Cell Markers, Neuroscience | |
| protein S100-A1, S100 alpha, S100 calcium binding protein A1, S100 calcium-binding protein A1S100, S-100 protein alpha chain, S-100 protein subunit alpha, S100 protein, alpha polypeptide, S100A, S100-alpha | |
| S100A1 | |
| IgG | |
| Affinity Purified | |
| This antibody reacts with cow S-100 protein a and b. It shows strong crossreactivity with human S-100 a and b. In the brain, it labels glial and ependymal cells. Melanocytes and Langerhans cells of the skin stain positive with this antibody. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title