missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A/B Antibody (8B10cc), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-8330
This item is not returnable.
View return policy
Description
S100A/B Monoclonal specifically detects S100A/B in Human samples. It is validated for Western Blot, Sandwich ELISA.
Specifications
| S100A/B | |
| Monoclonal | |
| Unconjugated | |
| P23297 | |
| S100A1 | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Human brain S100 proteins. | |
| 0.2 mg | |
| Astrocyte Markers, Breast Cancer, Cell Biology, Cytoskeleton Markers, Neuronal Cell Markers, Neuroscience | |
| 6271 | |
| Human | |
| Purified |
| Immunoassay, ELISA | |
| 8B10cc | |
| Western Blot 1:100-1:2000, Sandwich ELISA 1:100-1:2000 | |
| protein S100-A1, S100 alpha, S100 calcium binding protein A1, S100 calcium-binding protein A1S100, S-100 protein alpha chain, S-100 protein subunit alpha, S100 protein, alpha polypeptide, S100A, S100-alpha | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Specific to S-100 beta beta and alpha beta. | |
| Store at 4C. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction